siRNA Details
| Gene Symbol: Dctn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159565.1 | NP_001153037.1 | 3 | 242 | ||
| NM_016890.4 | NP_058586.3 | 3 | 242 | |||
| GenBank mRNA | AF098508.1 | - | 3 | 221 | ||
| AK002720.1 | - | 3 | 233 | |||
| AK003255.1 | - | 3 | 217 | |||
| AK011840.1 | - | 3 | 239 | |||
| AK159344.1 | - | 3 | 240 | |||
| AV493721.1 | - | 3 | 228 | |||
| BC061120.1 | - | 3 | 223 | 6774742 | ||
| BY141790.1 | - | 3 | 249 | |||

Dctn3
Genomic Map