siRNA Details
Gene Symbol: Cd164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016898.2 | NP_058594.1 | 2 | 262 | ||
GenBank mRNA | AB014464.1 | - | 2 | 227 | ||
AB028895.1 | - | 2 | 262 | |||
AF299345.1 | - | 2 | 227 | |||
AK006592.1 | - | 2 | 121 | |||
AK018009.1 | - | 2 | 262 | |||
AK145131.1 | - | 2 | 321 | |||
AK169275.1 | - | 2 | 302 | |||
AK194033.1 | - | 2 | 129 | |||
BC005414.1 | - | 2 | 250 | 3600535 |
