siRNA Details
| Gene Symbol: Hpcal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016677.4 | NP_057886.1 | 3 | 696 | ||
| GenBank mRNA | AK002849.1 | - | 3 | 679 | ||
| AK182666.1 | - | 1 | 88 | |||
| AK213639.1 | - | 1 | 88 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hpcal1| Gene Symbol: Hpcal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016677.4 | NP_057886.1 | 3 | 696 | ||
| GenBank mRNA | AK002849.1 | - | 3 | 679 | ||
| AK182666.1 | - | 1 | 88 | |||
| AK213639.1 | - | 1 | 88 | |||