siRNA Details
| Gene Symbol: Zpbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185153.1 | NP_001172082.1 | 5 | 748 | ||
| NM_015785.2 | NP_056600.1 | 5 | 919 | |||
| GenBank mRNA | AK006274.1 | - | 2 | 261 | ||
| AK018889.1 | - | 2 | 263 | |||
| AK018921.1 | - | 2 | 263 | |||
| AK029541.1 | - | 2 | 263 | |||
| BC139450.1 | - | 5 | 888 | 8862472 | ||
| BC139453.1 | - | 5 | 888 | 8862475 | ||
| BC145628.1 | - | 5 | 717 | 9054196 | ||
| D17569.1 | - | 5 | 908 | |||

Zpbp
Genomic Map