siRNA Details
| Gene Symbol: Zpbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185153.1 | NP_001172082.1 | 6, 7 | 836 | ||
| NM_015785.2 | NP_056600.1 | 6, 7 | 1007 | |||
| GenBank mRNA | AK006274.1 | - | 3, 4 | 349 | ||
| AK018889.1 | - | 3, 4 | 351 | |||
| AK018921.1 | - | 3, 4 | 351 | |||
| AK029541.1 | - | 3, 4 | 351 | |||
| BC139450.1 | - | 6, 7 | 976 | 8862472 | ||
| BC139453.1 | - | 6, 7 | 976 | 8862475 | ||
| BC145628.1 | - | 6, 7 | 805 | 9054196 | ||
| D17569.1 | - | 6, 7 | 996 | |||

Zpbp
Genomic Map