siRNA Details
| Gene Symbol: Vti1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016800.3 | NP_058080.2 | 3, 4 | 489 | ||
| GenBank mRNA | AF035208.1 | - | 3, 4 | 376 | ||
| AF035821.1 | - | 3, 4 | 362 | |||
| AK002289.1 | - | 3, 4 | 481 | |||
| AK160624.1 | - | 2, 3 | 423 | |||
| BC002290.1 | - | 2, 3 | 414 | |||
| BC009810.1 | - | 3, 4 | 419 | 4220215 | ||
| BC080204.1 | - | 3, 4 | 465 | 3672445 | ||
| CX239415.1 | - | 3, 4 | 466 | |||

Vti1b
Genomic Map