siRNA Details
| Gene Symbol: Blcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016916.3 | NP_058612.3 | 2 | 413 | ||
| GenBank mRNA | AB037891.1 | - | 2 | 368 | ||
| AK011673.1 | - | 2 | 390 | |||
| AK018127.1 | - | 2 | 443 | |||
| AK170338.1 | - | 2 | 390 | |||
| BC026935.1 | - | 2 | 377 | 3597231 | ||
| BC083339.1 | - | 2 | 377 | 6832329 | ||
| BM950751.1 | - | 2 | 354 | |||
| BY718283.1 | - | 2 | 443 | |||

Blcap
Genomic Map