siRNA Details
| Gene Symbol: Col5a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016919.2 | NP_058615.1 | 8 | 1158 | ||
| GenBank mRNA | AB040491.1 | - | 8 | 1162 | ||
| AF176645.1 | - | 8 | 1158 | |||
| BC138417.1 | - | 8 | 1123 | 8861437 | ||
| BC138419.1 | - | 8 | 1123 | 8861439 | ||
| BC145269.1 | - | 8 | 1120 | |||

Col5a3
Genomic Map