siRNA Details
| Gene Symbol: Rab11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017382.5 | NP_059078.2 | 4, 5 | 652 | ||
| GenBank mRNA | AB232605.1 | - | 4, 5 | 519 | ||
| AK014268.1 | - | 4, 5 | 653 | |||
| AK031143.1 | - | 4, 5 | 555 | |||
| AK132072.1 | - | 4, 5 | 636 | |||
| AK132159.1 | - | 4, 5 | 638 | |||
| AK147122.1 | - | 4, 5 | 653 | |||
| AK219017.1 | - | 1, 2 | 96 | |||
| BC010722.1 | - | 4, 5 | 641 | 2811973 | ||
| D50500.1 | - | 2, 3 | 160 | |||

Rab11a
Genomic Map