siRNA Details
| Gene Symbol: Rab11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017382.5 | NP_059078.2 | 4 | 631 | ||
| GenBank mRNA | AB232605.1 | - | 4 | 498 | ||
| AK014268.1 | - | 4 | 632 | |||
| AK031143.1 | - | 4 | 534 | |||
| AK132072.1 | - | 4 | 615 | |||
| AK132159.1 | - | 4 | 617 | |||
| AK147122.1 | - | 4 | 632 | |||
| BC010722.1 | - | 4 | 620 | 2811973 | ||
| D50500.1 | - | 2 | 139 | |||

Rab11a
Genomic Map