siRNA Details
| Gene Symbol: Rab11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017382.5 | NP_059078.2 | 2 | 274 | ||
| GenBank mRNA | AB232605.1 | - | 2 | 141 | ||
| AK014268.1 | - | 2 | 275 | |||
| AK031143.1 | - | 2 | 178 | |||
| AK132072.1 | - | 2 | 258 | |||
| AK132159.1 | - | 2 | 260 | |||
| AK147122.1 | - | 2 | 275 | |||
| BC010722.1 | - | 2 | 263 | 2811973 | ||

Rab11a
Genomic Map