siRNA Details
| Gene Symbol: Caprin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111289.1 | NP_001104759.1 | 7, 8 | 1015 | ||
| NM_001111290.1 | NP_001104760.1 | 7, 8 | 1015 | |||
| NM_001111291.1 | NP_001104761.1 | 8, 9 | 1055 | |||
| NM_001111292.1 | NP_001104762.1 | 7, 8 | 1015 | |||
| NM_016739.3 | NP_058019.2 | 8, 9 | 1055 | |||
| GenBank mRNA | AB373955.1 | - | 7, 8 | 877 | ||
| AK133891.1 | - | 8, 9 | 1017 | |||
| AK141992.1 | - | 7, 8 | 957 | |||
| AK147514.1 | - | 7, 8 | 2826 | |||
| AK148203.1 | - | 7, 8 | 1002 | |||
| AK157707.1 | - | 8, 9 | 1039 | |||
| AK161131.1 | - | 2, 3 | 150 | |||
| AK166461.1 | - | 7, 8 | 973 | |||
| AK168676.1 | - | 7, 8 | 1010 | |||
| AK170830.1 | - | 8, 9 | 1017 | |||
| AK187984.1 | - | 1, 2 | 45 | |||
| AK193449.1 | - | 1, 2 | 45 | |||
| AK200310.1 | - | 1, 2 | 45 | |||
| BC052427.1 | - | 8, 9 | 1034 | 6836041 | ||
| U27838.1 | - | 7, 8 | 992 | |||

Caprin1
Genomic Map