siRNA Details
| Gene Symbol: Ear7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017385.1 | NP_059081.1 | 1 | 268 | ||
| Gene Symbol: Ear6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053111.2 | NP_444341.1 | 2 | 335 | ||
| GenBank mRNA | AK157836.1 | - | 2 | 332 | ||
| AY316150.1 | - | 2 | 335 | |||
| BC147690.1 | - | 1 | 567 | 9056663 | ||
| BC147697.1 | - | 1 | 567 | 9056670 | ||

Ear7
Genomic Map