siRNA Details
| Gene Symbol: Svs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017390.4 | NP_059086.3 | 2 | 816 | ||
| GenBank mRNA | AK020661.1 | - | 2 | 260 | ||
| AK035204.1 | - | 2 | 741 | |||
| AK040724.1 | - | 2 | 742 | |||
| AK090151.1 | - | 2 | 738 | |||
| AK090302.1 | - | 2 | 742 | |||
| AK136679.1 | - | 2 | 740 | |||
| AK144285.1 | - | 2 | 741 | |||
| AK144389.1 | - | 2 | 741 | |||
| AK144429.1 | - | 2 | 741 | |||
| AK144524.1 | - | 2 | 738 | |||
| AK165704.1 | - | 2 | 741 | |||
| AK165713.1 | - | 2 | 741 | |||
| AK165727.1 | - | 2 | 738 | |||
| AK165753.1 | - | 2 | 741 | |||
| AK165825.1 | - | 2 | 741 | |||
| BC107275.1 | - | 2 | 741 | 40052650 | ||
| BC107276.1 | - | 2 | 741 | |||

Svs2
Genomic Map