siRNA Details
| Gene Symbol: Slc5a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017391.3 | NP_059087.2 | 2 | 1466 | ||
| GenBank mRNA | AK133628.1 | - | 1 | 2672 | ||
| AK144718.1 | - | 2 | 1467 | |||
| BC140982.1 | - | 1 | 1013 | 9055594 | ||
| BC140983.1 | - | 1 | 1013 | 9055595 | ||

Slc5a3
Genomic Map