siRNA Details
| Gene Symbol: Slc5a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017391.3 | NP_059087.2 | 2 | 2468 | ||
| GenBank mRNA | BC140982.1 | - | 1 | 2015 | 9055594 | |
| BC140983.1 | - | 1 | 2015 | 9055595 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc5a3| Gene Symbol: Slc5a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017391.3 | NP_059087.2 | 2 | 2468 | ||
| GenBank mRNA | BC140982.1 | - | 1 | 2015 | 9055594 | |
| BC140983.1 | - | 1 | 2015 | 9055595 | ||