siRNA Details
| Gene Symbol: Ppm1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016910.3 | NP_058606.3 | 3 | 982 | ||
| GenBank mRNA | AF200464.1 | - | 3 | 763 | ||
| AK157818.1 | - | 3 | 970 | |||
| AK171714.1 | - | 3 | 962 | |||
| BC023492.1 | - | 3 | 858 | 5026376 | ||
| BC051966.1 | - | 3 | 982 | 5689560 | ||

Ppm1d
Genomic Map