siRNA Details
| Gene Symbol: Gal3st1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177691.1 | NP_001171162.1 | 3 | 1206 | ||
| NM_001177703.1 | NP_001171174.1 | 3 | 1337 | |||
| NM_016922.3 | NP_058618.2 | 4 | 1555 | |||
| GenBank mRNA | AB032939.1 | - | 2 | 1132 | ||
| AK002510.1 | - | 3 | 1202 | |||
| AK007645.1 | - | 3 | 1337 | |||
| BC026806.1 | - | 4 | 1478 | 4215616 | ||

Gal3st1
Genomic Map