siRNA Details
| Gene Symbol: Ngef | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111314.1 | NP_001104784.1 | 9 | 1557 | ||
| NM_019867.2 | NP_063920.2 | 7 | 1194 | |||
| GenBank mRNA | AJ238898.1 | - | 7 | 1175 | ||
| AK138433.1 | - | 7 | 1163 | |||
| AY038025.1 | - | 7 | 1169 | |||
| BC039279.1 | - | 9 | 1557 | 4973826 | ||

Ngef
Genomic Map