siRNA Details
| Gene Symbol: Rps28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC010987.1 | - | 4 | 283 | 4218716 | |
| BC090982.1 | - | 4 | 281 | 30316901 | ||
| DQ566373.1 | - | 17 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rps28| Gene Symbol: Rps28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC010987.1 | - | 4 | 283 | 4218716 | |
| BC090982.1 | - | 4 | 281 | 30316901 | ||
| DQ566373.1 | - | 17 | ||||