siRNA Details
| Gene Symbol: Actr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016860.1 | NP_058556.1 | 4 | 299 | ||
| GenBank mRNA | AB010297.1 | - | 4 | 299 | ||
| AK010632.1 | - | 4 | 296 | |||
| AK019518.1 | - | 4 | 303 | |||
| AK088062.1 | - | 4 | 306 | |||
| AK162248.1 | - | 4 | 314 | |||
| AK167450.1 | - | 4 | 306 | |||
| AK168889.1 | - | 4 | 303 | |||
| BC007131.1 | - | 4 | 314 | 3599656 | ||

Actr1a
Genomic Map