siRNA Details
| Gene Symbol: Atxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016843.3 | NP_058539.2 | 10 | 1303 | ||
| GenBank mRNA | AK002504.1 | - | 10 | 1414 | ||
| AK003530.1 | - | 10 | 1230 | |||
| AK047170.1 | - | 10 | 1353 | |||
| AK049706.1 | - | 10 | 1347 | |||
| AK077738.1 | - | 10 | 1306 | |||
| AK078459.1 | - | 10 | 1383 | |||
| AK153271.1 | - | 10 | 1305 | |||
| AK161950.1 | - | 1320 | ||||
| AK165941.1 | - | 10 | 1328 | |||
| AK166899.1 | - | 10 | 1388 | |||
| AK170507.1 | - | 10 | 1320 | |||
| AK170751.1 | - | 10 | 1329 | |||
| AK185054.1 | - | 3 | 280 | |||
| BC002285.1 | - | 4 | 472 | |||
| BC016410.1 | - | 10 | 1313 | 3968016 | ||
| BC046802.1 | - | 10 | 1439 | 6404607 | ||

Atxn10
Genomic Map