siRNA Details
| Gene Symbol: Dnal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017470.2 | NP_059498.2 | 4 | 418 | ||
| GenBank mRNA | AB010031.1 | - | 4 | 396 | ||
| AK002657.1 | - | 4 | 369 | |||
| AK029618.1 | - | 3 | 229 | |||
| AK132061.1 | - | 4 | 408 | |||
| BC005426.1 | - | 4 | 347 | 3593932 | ||

Dnal4
Genomic Map