siRNA Details
| Gene Symbol: Sult2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017465.2 | NP_059493.2 | 1, 2 | 175 | ||
| GenBank mRNA | AF026072.1 | - | 1, 2 | 142 | ||
| AF478566.1 | - | 1 | 298 | |||
| BB867946.1 | - | 1, 2 | 176 | |||
| BC009811.1 | - | 1, 2 | 217 | 4008307 | ||
| BC009813.1 | - | 1, 2 | 105 | 4039050 | ||

Sult2b1
Genomic Map