siRNA Details
| Gene Symbol: Sult2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017465.2 | NP_059493.2 | 3 | 481 | ||
| GenBank mRNA | AF026072.1 | - | 3 | 448 | ||
| BC009811.1 | - | 3 | 523 | 4008307 | ||
| BC009813.1 | - | 3 | 411 | 4039050 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult2b1| Gene Symbol: Sult2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017465.2 | NP_059493.2 | 3 | 481 | ||
| GenBank mRNA | AF026072.1 | - | 3 | 448 | ||
| BC009811.1 | - | 3 | 523 | 4008307 | ||
| BC009813.1 | - | 3 | 411 | 4039050 | ||