siRNA Details
| Gene Symbol: Cd160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163496.1 | NP_001156968.1 | 3 | 298 | ||
| NM_001163497.1 | NP_001156969.1 | 3 | 298 | |||
| NM_018767.3 | NP_061237.3 | 3 | 221 | |||
| GenBank mRNA | AF060982.1 | - | 3 | 192 | ||
| AK042093.1 | - | 3 | 222 | |||
| AK139789.1 | - | 3 | 299 | |||
| AK145332.1 | - | 3 | 279 | |||
| BC021596.1 | - | 3 | 193 | 5098004 | ||
| DV660330.1 | - | 3 | 254 | |||

Cd160
Genomic Map