siRNA Details
| Gene Symbol: B3galt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019420.2 | NP_062293.1 | 1 | 1300 | ||
| GenBank mRNA | AF082504.1 | - | 1 | 1293 | ||
| AK007600.1 | - | 1 | 1288 | |||
| AK166333.1 | - | 1 | 1257 | |||
| KJ650213.1 | - | 1 | 1074 | |||

B3galt4
Genomic Map