siRNA Details
| Gene Symbol: Zfp326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018759.2 | NP_061229.2 | 6 | 889 | ||
| GenBank mRNA | AB012725.1 | - | 6 | 855 | ||
| AK017693.1 | - | 6 | 890 | |||
| AK146438.1 | - | 6 | 870 | |||
| BC005567.1 | - | 6 | 880 | |||
| BC037055.1 | - | 4 | 430 | 4948371 | ||
| BC039041.1 | - | 5 | 2894 | |||
| BC042769.1 | - | 1 | 1785 | |||
| BU503985.1 | - | 3 | 273 | |||

Zfp326
Genomic Map