siRNA Details
| Gene Symbol: Zfp326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018759.2 | NP_061229.2 | 7 | 999 | ||
| GenBank mRNA | AB012725.1 | - | 7 | 965 | ||
| AK017693.1 | - | 7 | 1000 | |||
| BC037055.1 | - | 5 | 540 | 4948371 | ||
| BC039041.1 | - | 6 | 3004 | |||
| BU503985.1 | - | 4 | 383 | |||

Zfp326
Genomic Map