siRNA Details
| Gene Symbol: Chst2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018763.2 | NP_061233.2 | 2 | 1864 | ||
| GenBank mRNA | AB011451.1 | - | 2 | 1316 | ||
| AB011452.1 | - | 2 | 1316 | |||
| BC051963.1 | - | 2 | 1251 | 5707543 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chst2| Gene Symbol: Chst2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018763.2 | NP_061233.2 | 2 | 1864 | ||
| GenBank mRNA | AB011451.1 | - | 2 | 1316 | ||
| AB011452.1 | - | 2 | 1316 | |||
| BC051963.1 | - | 2 | 1251 | 5707543 | ||