siRNA Details
| Gene Symbol: Prss16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019429.2 | NP_062302.1 | 11, 12 | 1525 | ||
| GenBank mRNA | AJ131775.1 | - | 11, 12 | 1525 | ||
| AK040376.1 | - | 4, 5 | 908 | |||
| AK088019.1 | - | 11, 12 | 1553 | |||
| BC125481.1 | - | 11, 12 | 1523 | 40129996 | ||
| BC131955.1 | - | 11, 12 | 1523 | 40130232 | ||

Prss16
Genomic Map