siRNA Details
| Gene Symbol: Mtmr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040699.1 | NP_001035789.1 | 4 | 520 | ||
| GenBank mRNA | AK028160.1 | - | 4 | 485 | ||
| AK081973.1 | - | 4 | 506 | |||
| AK148768.1 | - | 4 | 521 | |||
| BC032254.1 | - | 4 | 522 | 5368816 | ||
| BY741806.1 | - | 4 | 521 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001107312, XM_006253195.

Mtmr7
Genomic Map