siRNA Details
| Gene Symbol: Crlf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277106.1 | NP_001264035.1 | 3 | 469 | ||
| NM_018776.2 | NP_061246.1 | 3 | 469 | |||
| GenBank mRNA | AF046060.1 | - | 3 | 444 | ||
| AF120152.1 | - | 3 | 378 | |||
| AK037880.1 | - | 3 | 417 | |||
| AK078569.1 | - | 3 | 414 | |||
| AK137528.1 | - | 3 | 421 | |||
| AK146798.1 | - | 3 | 417 | |||
| BC004780.1 | - | 3 | 416 | 3257685 | ||

Crlf3
Genomic Map