siRNA Details
| Gene Symbol: Stk19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019442.3 | NP_062315.1 | 3 | 285 | ||
| GenBank mRNA | AF099934.1 | - | 3 | 219 | ||
| AK146739.1 | - | 3 | 287 | |||
| AK168865.1 | - | 3 | 287 | |||
| AK215314.1 | - | 2 | 45 | |||
| AK218509.1 | - | 2 | 114 | |||

Stk19
Genomic Map