siRNA Details
| Gene Symbol: KRT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005556.3 | NP_005547.3 | 9 | 1646 | ||
| XM_017019294.1 | XP_016874783.1 | 9 | 1372 | |||
| GenBank mRNA | AK025881.1 | - | 9 | 1573 | ||
| AK128505.1 | - | 8 | 4314 | |||
| AK128848.1 | - | 10 | 1501 | |||
| AK129924.1 | - | 10 | 1580 | |||
| BC002700.2 | - | 9 | 1561 | 3610347 | ||
| BC009521.1 | - | 5 | 733 | |||

KRT7
Genomic Map