siRNA Details
| Gene Symbol: Cldn8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018778.3 | NP_061248.1 | 1 | 254 | ||
| GenBank mRNA | AF087826.1 | - | 1 | 106 | ||
| AK133810.1 | - | 1 | 254 | |||
| BC003868.1 | - | 1 | 206 | 3490218 | ||
 Back To Top
 Back To TopMore Information
Other Species Targeted
 This siRNA also targets the following transcripts in Rat: NM_001037774, BC099804.
					            	This siRNA also targets the following transcripts in Rat: NM_001037774, BC099804.Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Cldn8
Cldn8 Genomic Map
Genomic Map Back To Top
 Back To Top