siRNA Details
| Gene Symbol: Unc93b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161428.1 | NP_001154900.1 | 9 | 1179 | ||
| NM_019449.2 | NP_062322.2 | 10 | 1453 | |||
| GenBank mRNA | AK030488.1 | - | 10 | 1413 | ||
| AK155556.1 | - | 10 | 1411 | |||
| AK165060.1 | - | 10 | 1413 | |||
| BC018388.1 | - | 10 | 1420 | 4209296 | ||
| BC023731.1 | - | 4 | 878 | 5352340 | ||
| BC025587.1 | - | 3 | 355 | |||

Unc93b1
Genomic Map