siRNA Details
| Gene Symbol: Unc93b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161428.1 | NP_001154900.1 | 3 | 408 | ||
| NM_019449.2 | NP_062322.2 | 3 | 408 | |||
| GenBank mRNA | AK030488.1 | - | 3 | 369 | ||
| AK039526.1 | - | 3 | 369 | |||
| AK155556.1 | - | 3 | 366 | |||
| AK162535.1 | - | 3 | 361 | |||
| AK165060.1 | - | 3 | 368 | |||
| BC018388.1 | - | 3 | 375 | 4209296 | ||

Unc93b1
Genomic Map