siRNA Details
| Gene Symbol: Nfat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286260.1 | NP_001273189.1 | 5 | 1360 | ||
| NM_018823.2 | NP_061293.2 | 6 | 1414 | |||
| NM_133957.3 | NP_598718.2 | 7 | 1476 | |||
| GenBank mRNA | AF162853.1 | - | 5 | 1271 | ||
| AF200687.1 | - | 6 | 1049 | |||
| AF228705.1 | - | 3 | 1114 | |||
| AF228706.1 | - | 3 | 1114 | |||
| AF369980.1 | - | 7 | 1115 | |||
| AF453571.2 | - | 7 | 1084 | |||
| AK134022.1 | - | 7 | 1480 | |||

Nfat5
Genomic Map