siRNA Details
| Gene Symbol: Tollip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023764.3 | NP_076253.1 | 3 | 349 | ||
| GenBank mRNA | AJ242971.1 | - | 3 | 353 | ||
| AK015062.1 | - | 292 | ||||
| AK016482.1 | - | 3 | 350 | |||
| AK045422.1 | - | 3 | 333 | |||
| AK049263.1 | - | 3 | 371 | |||
| AK049394.1 | - | 3 | 295 | |||
| AK078391.1 | - | 359 | ||||
| AK161312.1 | - | 3 | 336 | |||
| AK189903.1 | - | 2 | 123 | |||
| BC053515.1 | - | 3 | 336 | |||
| BC062139.1 | - | 3 | 330 | 6334946 | ||

Tollip
Genomic Map