siRNA Details
| Gene Symbol: Mkrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018810.2 | NP_061280.2 | 3 | 539 | ||
| GenBank mRNA | AB026986.1 | - | 3 | 644 | ||
| AF192785.1 | - | 3 | 564 | |||
| AK005137.1 | - | 3 | 373 | |||
| AK029500.1 | - | 3 | 579 | |||
| AK077060.1 | - | 3 | 540 | |||
| AK078962.1 | - | 3 | 469 | |||
| AK186489.1 | - | 1 | 147 | |||
| AK207305.1 | - | 1 | 147 | |||
| AK210725.1 | - | 1 | 147 | |||
| BC003329.1 | - | 3 | 528 | 3158135 | ||
| Gene Symbol: Mkrn1-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF494488.1 | - | LIT1345 Makorin1-p1 |
1 | 352 | |

Mkrn1
Genomic Map