siRNA Details
| Gene Symbol: Nup210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018815.2 | NP_061285.2 | 11 | 1461 | ||
| GenBank mRNA | AF113751.1 | - | 11 | 1429 | ||
| AK157272.1 | - | 11 | 1453 | |||
| AK173056.1 | - | 11 | 1415 | |||
| BC052468.1 | - | 11 | 1454 | 6835038 | ||

Nup210
Genomic Map