siRNA Details
| Gene Symbol: Nup210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018815.2 | NP_061285.2 | 9 | 1150 | ||
| GenBank mRNA | AF113751.1 | - | 9 | 1118 | ||
| AK157272.1 | - | 9 | 1142 | |||
| AK173056.1 | - | 9 | 1104 | |||
| BC052468.1 | - | 9 | 1143 | 6835038 | ||

Nup210
Genomic Map