siRNA Details
| Gene Symbol: Pcnx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018814.3 | NP_061284.2 | 30 | 5851 | ||
| GenBank mRNA | AF096286.1 | - | 24 | 3681 | ||
| AK014387.1 | - | 4 | 539 | |||
| AK129221.1 | - | 25 | 3731 | |||
| AK154122.1 | - | 11 | 1579 | |||
| AK170320.1 | - | 11 | 1579 | |||
| BC057333.1 | - | 23 | 3594 | |||
| BC066849.1 | - | 25 | 3516 | |||
| BC098370.1 | - | 25 | 3441 | |||

Pcnx
Genomic Map