siRNA Details
| Gene Symbol: Pcnx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018814.3 | NP_061284.2 | 6 | 2156 | ||
| GenBank mRNA | AF096286.1 | - | 1 | 186 | ||
| AK129221.1 | - | 1 | 36 | |||
| BC057333.1 | - | 1 | 232 | |||
| CN535767.1 | - | 1 | 344 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcnx| Gene Symbol: Pcnx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018814.3 | NP_061284.2 | 6 | 2156 | ||
| GenBank mRNA | AF096286.1 | - | 1 | 186 | ||
| AK129221.1 | - | 1 | 36 | |||
| BC057333.1 | - | 1 | 232 | |||
| CN535767.1 | - | 1 | 344 | |||