siRNA Details
| Gene Symbol: Tbc1d8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018775.4 | NP_061245.3 | 15 | 2644 | ||
| GenBank mRNA | AK138630.1 | - | 15 | 2642 | ||
| AK141520.1 | - | 15 | 2633 | |||
| AK141816.1 | - | 15 | 2598 | |||
| BC005421.1 | - | 12 | 1979 | |||
| BC065081.1 | - | 15 | 2578 | 6832495 | ||
| U88873.1 | - | 11 | 1864 | |||

Tbc1d8
Genomic Map