siRNA Details
| Gene Symbol: Tbc1d8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018775.4 | NP_061245.3 | 16 | 2729 | ||
| GenBank mRNA | AK138630.1 | - | 16 | 2727 | ||
| AK141520.1 | - | 16 | 2718 | |||
| AK141816.1 | - | 16 | 2683 | |||
| AK162371.1 | - | 10 | 1504 | |||
| BC005421.1 | - | 13 | 2064 | |||
| BC065081.1 | - | 16 | 2663 | 6832495 | ||
| U88873.1 | - | 12 | 1949 | |||

Tbc1d8
Genomic Map