siRNA Details
| Gene Symbol: Pde3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018779.1 | NP_061249.1 | 5 | 1777 | ||
| GenBank mRNA | AF099187.1 | - | 5 | 1777 | ||
| AK004638.1 | - | 5 | 1082 | |||
| AK052374.1 | - | 5 | 1055 | |||
| AK164864.1 | - | 5 | 558 | |||
| BC138265.1 | - | 5 | 1735 | 8861284 | ||
| BC138266.1 | - | 5 | 1735 | 8861285 | ||

Pde3a
Genomic Map