siRNA Details
| Gene Symbol: Extl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018788.3 | NP_061258.2 | 7 | 3273 | ||
| GenBank mRNA | AF083550.1 | - | 5 | 3063 | ||
| AK137842.1 | - | 5 | 2350 | |||
| AK220344.1 | - | 5 | 2616 | |||
| BC058526.1 | - | 5 | 2664 | |||
| BC065073.1 | - | 5 | 2738 | 6834580 | ||

Extl3
Genomic Map