siRNA Details
| Gene Symbol: Pqbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252528.1 | NP_001239457.1 | 4 | 331 | ||
| NM_001252529.1 | NP_001239458.1 | 4 | 397 | |||
| NM_019478.4 | NP_062351.2 | 4 | 526 | |||
| GenBank mRNA | AJ250406.1 | - | 3 | 277 | ||
| AK077652.1 | - | 4 | 356 | |||
| BC009657.1 | - | 3 | 299 | 3484619 | ||
| BC051673.1 | - | 4 | 336 | 30015428 | ||
| BC157976.1 | - | 4 | 431 | 9088137 | ||
| BQ550428.1 | - | 4 | 402 | |||
| CJ143105.1 | - | 4 | 332 | |||
| CN718083.1 | - | 3 | 311 | |||

Pqbp1
Genomic Map